missing translation for 'onlineSavingsMsg'
Learn More

CD249 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA552138

Additional Details : Weight : 0.02000kg

Product Code. 15740091

  • £399.00 / 100µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: SHLPSSTASP SGPPAQDQDI CPASEDESGQ WKNFRLPDFV NPVHYDLHVK PLLEEDTYTG TVSISINLSA PTRYLWLHLR ETRITRLPEL KRPSGDQVQV RRCFEYKKQE YVVVE Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.

CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.
Specifications

Specifications

CD249
Polyclonal
Unconjugated
Enpep
6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51
Rabbit
Antigen affinity chromatography
RUO
2028
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q07075
Enpep
Recombinant protein corresponding to Human CD249
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only