Learn More
CD249 (BP-1) Monoclonal Antibody (6C3), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 12-5891-81
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The 6C3 monoclonal antibody reacts with mouse BP-1, also known as Ly-51 and 6C3 antigen. BP-1 is a 140 kDa type II transmembrane protein that is present on the surface as a disulfide-linked homodimer. This molecule is an enzyme with aminopeptidase A (APA) activity and is expressed by bone marrow stromal cells, thymic cortical epithelial cells and at early stages of mouse B cell development, on pre-B cells. The low level of BP-1 expression on pre-B cells can be upregulated in the presence of IL-7. Applications Reported: The 6C3 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 6C3 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.Specifications
CD249 (BP-1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P16406 | |
Enpep | |
Affinity chromatography | |
RUO | |
13809 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
6C3 | |
PE | |
Enpep | |
6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51 | |
Rat | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.