Learn More
Invitrogen™ CD249 (BP-1) Monoclonal Antibody (6C3), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£69.50 - £249.00
Specifications
| Antigen | CD249 (BP-1) |
|---|---|
| Clone | 6C3 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15299899
|
Invitrogen™
13-5891-81 |
50 μg |
£69.50
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15219909
|
Invitrogen™
13-5891-85 |
500 μg |
£249.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The 6C3 monoclonal antibody reacts with mouse BP-1, also known as Ly-51 and 6C3 antigen. BP-1 is a 140 kDa type II transmembrane protein that is present on the surface as a disulfide-linked homodimer. This molecule is an enzyme with aminopeptidase A (APA) activity and is expressed by bone marrow stromal cells, thymic cortical epithelial cells and at early stages of mouse B cell development, on pre-B cells. The low level of BP-1 expression on pre-B cells can be upregulated in the presence of IL-7. Applications Reported: The 6C3 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 6C3 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.Specifications
| CD249 (BP-1) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P16406 | |
| Enpep | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Enpep |
| 6C3 | |
| Flow Cytometry | |
| Biotin | |
| Rat | |
| Mouse | |
| 13809 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.