missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD248 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588715

Product Code. 16586851

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QDPWAAEPRA ACGPSSCYAL FPRRRTFLEA WRACRELGGD LATPRTPEEA QRVDSLVGAG PASRLLWIGL QRQARQCQLQ RPLRGFTWTT GDQDTAFTNW AQPASGGPCP AQRCVALEAS GEHRWLEGSC TLA; Positive Samples: A-375, Mouse spleen, Mouse thymus; Cellular Location: Membrane, Single-pass type I membrane protein.

CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD248
Polyclonal
Unconjugated
CD248
2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1
Rabbit
Affinity Chromatography
RUO
293669, 57124, 70445
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.36 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q91V98, Q9HCU0
CD248
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.