missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD244 Monoclonal Antibody (eBioC1.7 (C1.7)), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD244 |
|---|---|
| Clone | eBioC1.7 (C1.7) |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089377
|
Invitrogen™
H080T02Y04-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089047
|
Invitrogen™
H080T03Y04-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD244 Monoclonal antibody specifically detects CD244 in Human samples. It is validated for Flow CytometrySpecifications
| CD244 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 2B4; C9.1; CD244; CD244 long isoform; CD244 molecule; Cd244 molecule, natural killer cell receptor 2B4; CD244 natural killer cell receptor 2B4; CD244 short isoform; F730046O15Rik; h2B4; Ly90; NAIL; natural killer cell receptor 2B4; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell receptor 2B4; NK cell type I receptor protein 2B4; NKR Nmrk; NKR2B4; Nmrk; non MHC restricted killing associated; non-MHC restricted killing associated; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4; transmembrane NK cell receptor 2B4 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| eBioC1.7 (C1.7) | |
| Flow Cytometry | |
| NovaFluor Yellow 660 | |
| Mouse | |
| Human | |
| Q9BZW8 | |
| 51744 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD244 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title