missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Rat, anti-Mouse, FITC, Clone: M1/69, STEMCELL Technologies
Rat Monoclonal Antibody
POA
Specifications
| Antigen | CD24 |
|---|---|
| Clone | M1/69 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
Description
- The M1/69 antibody reacts with murine CD24, an ∼35 - 45 kDa glycoprotein expressed on lymphocytes, granulocytes, thymocytes, erythrocytes, epithelial cells, neurons, and dendritic cells
- CD24 is differentially expressed during T and B cell development, and is not found on the majority of mature peripheral T cells or plasma cells
- It is expressed on pro-B, pre-B and mature B cells, but expression is strongly down-regulated after B cell activation
- CD24 is involved in regulating B cell apoptosis and in preventing terminal differentiation of B cells into antibody-secreting plasmablasts and plasma cells
- It acts as an adhesion or co-stimulatory molecule and is involved in lymphocyte activation, proliferation and differentiation through homophilic interactions or by binding to ligands such as P-selectin (CD62P)
Specifications
| CD24 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Mouse | |
| Primary |
| M1/69 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title