missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ CD24 (GPI-Linked Surface Mucin) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures.

Brand:  Epredia™ MS-1279-P1ABX

 View more versions of this product

Product Code. 12609456

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.

Host Species: Mouse

Clone: SN3b

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line

Molecular Weight: 30-70 kDa

Positive Control: Breast CA, Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: IVD; Int'l: IVD

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD24 (GPI-Linked Surface Mucin) Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
B-Cell
Human
IgM κ
Flow Cytometry, Immunohistochemistry (Paraffin)
SN3b
Unconjugated
Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.