Learn More
CD238 Mouse anti-Human, BUV615, Clone: BRIC 203, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 751404
Product Code. 17202585
  • ONLINE EXCLUSIVE PRICE
    £586.00
    £578.66 / 50µg
    Save £7.34
    1% Off
Description
CD238 Monoclonal antibody specifically detects CD238 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD238
Monoclonal
Brilliant Ultraviolet 615
KEL; KELL; Kell blood group, metallo-endopeptidase; ECE3
Human erythrocytes
RUO
Human
IgG1 κ
Flow Cytometry
BRIC 203
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.