missing translation for 'onlineSavingsMsg'
Learn More

CD235a Mouse anti-Human, PE, Clone: GA-R2 (HIR2), BD

Mouse Monoclonal Antibody

£117.21 - £236.75

Specifications

Antigen CD235a
Clone GA-R2 (HIR2)
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15890719
View Documents
BD Biosciences
561051
25 μg
ONLINE EXCLUSIVE PRICE
£120.00
£117.21 / Each
Save £2.79
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15828278
View Documents
BD Biosciences
555570
0.1 mg
ONLINE EXCLUSIVE PRICE
£426.00
£236.75 / Each
Save £189.25
44% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The GA-R2 (also known as HIR2) monoclonal antibody specifically binds to CD235a, otherwise known as Glycophorin A (GYPA, GPA), PAS-2, MN sialoglycoprotein (MNS, MN), and Sialoglycoprotein A/alpha. CD235a is a type I transmembrane sialoglycoprotein that is expressed on human erythrocytes, erythroid precursor cells and certain leukemic cell types. This antibody is useful for the identification and characterization of erythrocytes, certain myeloid leukemic cell types, and studies of erythroid cell development and infectious diseases with erythrocyte involvement. Mature, non-nucleated red blood cells characteristically express very high levels of glycophorin A.

Specifications

Specifications

CD235a
0.2mg/mL
Monoclonal
Mouse
Human
IgG2b κ
Affinity Purified
Glycophorin-A; GYPA; GPA; PAS-2; MN sialoglycoprotein; Sialoglycoprotein A
GA-R2 (HIR2)
Flow Cytometry
PE
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.