Learn More
Invitrogen™ CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), eBioscience™

Description
Description: The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells. Applications Reported: HIR2 can be used in flow cytometric analysis. Applications Tested: The HIR2 (also GA-R2) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and cell agglutination assay and can be used at approximately 0.1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD235a (Glycophorin A) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HIR2 (GA-R2) |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | GYPA |
| Gene Accession No. | P02724 |
| Gene Alias | AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.