Learn More
CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 64-9987-41
Description
Description: The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells. Applications Reported: This HIR2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIR2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No.
00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. Glycophorins A (GPA) and B (GPB) which are single pass membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation.
Specifications
| CD235a (Glycophorin A) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P02724 | |
| GYPA | |
| Affinity chromatography | |
| RUO | |
| 2993 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| HIR2 (GA-R2) | |
| Super Bright 645 | |
| GYPA | |
| CD235a; erythroid-lineage-specific membrane sialoglycoprotein; Glycophorin A; glycophorin A (MN blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; Glycophorin-A; Mi.V glycoprotein (24 AA); MN sialoglycoprotein; PAS-2; recombit glycophorin A-B Miltenberger-DR; Sialoglycoprotein alpha | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.