missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), PE-eFluor™ 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £408.00
Specifications
| Antigen | CD235a (Glycophorin A) |
|---|---|
| Clone | HIR2 (GA-R2) |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD235a (Glycophorins A, GPA) is a single pass membrane sialoglycoprotein expressed in mature erythrocytes and erythroid precursor cells. CD235a is the carrier of blood group M and N specificities. CD235a provides cells with a large mucin-like that may serve as a barrier to cell fusion, minimizing aggregation between red blood cells in the circulation. CD235a has been shown to act as a receptor for Sandei virus, parvovirus, and Hsa, an Streptococcus adhesin.Specifications
| CD235a (Glycophorin A) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| 2993 | |
| IgG2b κ | |
| Affinity chromatography | |
| GYPA |
| HIR2 (GA-R2) | |
| Flow Cytometry | |
| PE-eFluor 610 | |
| Mouse | |
| Human | |
| P02724 | |
| GYPA | |
| Primary | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title