missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD235a (Glycophorin A) Monoclonal Antibody (10F7MN), NovaFluor™ Blue 510, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£154.00 - £340.00

Specifications

Antigen CD235a (Glycophorin A)
Clone 10F7MN
Concentration 0.8 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055253
View Documents
Invitrogen™
H068T03B01-A
100 Tests
£340.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055457
View Documents
Invitrogen™
H068T02B01-A
25 Tests
£154.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD235a (Glycophorin A) Monoclonal antibody specifically detects CD235a (Glycophorin A) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD235a (Glycophorin A)
0.8 μg/Test
Monoclonal
Mouse
Human
P02724
2993
Primary
4°C, store in dark, DO NOT FREEZE!
10F7MN
Flow Cytometry
NovaFluor Blue 510
RUO
PBS with 0.09% sodium azide; pH 7.2
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha
IgG1 κ
Affinity chromatography
GYPA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.