missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23 Monoclonal Antibody (LVNC-4), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112065
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.Specifications
CD23 | |
Monoclonal | |
PE | |
FCER2 | |
BLAST-2; CD23; CD23 antigen; CD23A; CLEC4J; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc receptor alpha multiple ligand receptor; Fc receptor, IgE, low affinity II, alpha polypeptide; Fcalpha muR; Fcalpha RII; Fce2; fc-epsilon-RII; FCER2; Fcer2a; FceRII; IGEBF; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Ly-42; lymphocyte IgE receptor | |
Mouse | |
Protein A/G | |
RUO | |
2208 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
LVNC-4 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P06734 | |
FCER2 | |
White blood cells. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |