missing translation for 'onlineSavingsMsg'
Learn More

CD23/Fc epsilon RII Antibody (MM0164-2C23), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18418376 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418376 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418376 Supplier Novus Biologicals Supplier No. NBP212152AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD23/Fc epsilon RII
Applications Western Blot, Flow Cytometry, CyTOF, Neutralization
Classification Monoclonal
Clone MM0164-2C23
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Host Species Mouse
Immunogen Human recombinant CD23/Fc epsilon RII (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2208
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.