missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Rabbit anti-Mouse, Clone: 17, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90618-100UL
This item is not returnable.
View return policy
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Mouse samples. It is validated for ELISA.
Specifications
| CD229/SLAMF3/Lymphocyte Antigen 9 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4063 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 17 | |
| ELISA 1:5000-1:10000 | |
| CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
| Recombinant Mouse CD229/SLAMF3/Lymphocyte Antigen 9 protein | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction