missing translation for 'onlineSavingsMsg'
Learn More

CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (HLy9.1.25), DyLight 755, Novus Biologicals™

Product Code. 18432324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432324 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432324 Supplier Novus Biologicals Supplier No. NBP142422IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal antibody specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD229/SLAMF3/Lymphocyte Antigen 9
Applications Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF
Classification Monoclonal
Clone HLy9.1.25
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Host Species Mouse
Immunogen Mouse pre-B cell line 300.19 transfected with CD229/SLAMF3/Lymphocyte Antigen 9 cDNA
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4063
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.