missing translation for 'onlineSavingsMsg'
Learn More

CD229/SLAMF3/Lymphocyte Antigen 9 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18429786 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429786 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429786 Supplier Novus Biologicals Supplier No. NBP176557AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD229/SLAMF3/Lymphocyte Antigen 9 Polyclonal antibody specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD229/SLAMF3/Lymphocyte Antigen 9
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SLAMF3. The immunogen is located within amino acids 520 - 570 of SLAMF3. Amino Acid Squence: QPTPTSDSSSDSNLTTEED
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4063
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.