missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD229 Recombinant Rabbit Monoclonal Antibody (016), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£74.50 - £359.00
Specifications
| Antigen | CD229 |
|---|---|
| Clone | 016 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019700
![]() |
Invitrogen™
MA552400 |
100 μL |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019307
|
Invitrogen™
MA552401 |
20 μL |
£74.50
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD229 Recombinant Monoclonal antibody specifically detects CD229 in Mouse samples. It is validated for ELISASpecifications
| CD229 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
| Recombinant Mouse CD229 Protein (Lys 48-Arg453). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ly9 |
| 016 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q01965 | |
| 17085 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title