missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229 Mouse anti-Mouse , Alexa Fluor 647, Clone: Ly9.7.144, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566689
This item is not returnable.
View return policy
Description
CD229 Monoclonal antibody specifically detects CD229 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD229 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Lgp100; Lymphocyte antigen 9; Ly-9; Ly9; SLAM family member 3; SLAMF3; T100 | |
| Mouse Ly9 Transfected Cell Line | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| Ly9.7.144 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction