missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD223 (LAG-3) Monoclonal Antibody (3DS223H), Super Bright™ 702, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£191.00 - £360.00

Specifications

Antigen CD223 (LAG-3)
Clone 3DS223H
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16311654
View Documents
Invitrogen™
67-2239-41
25 Tests
£191.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16321654
GREENER_CHOICE
View Documents
Invitrogen™
67-2239-42
100 Tests
£360.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mouse Monoclonal Antibody

LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
Specifications

Specifications

CD223 (LAG-3)
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3
LAG3
IgG1 κ
Affinity chromatography
Antibody
3DS223H
Flow Cytometry
Super Bright 702
Mouse
Human
P18627
3902
hCD223-FC PFA 4%.
Primary
4°C, store in dark, DO NOT FREEZE!
LAG3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.