missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD22 Recombinant Rabbit Monoclonal Antibody (3F2M10)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558068

 View more versions of this product

Product Code. 30283398

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DSSKWVFEHP ETLYAWEGA DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSK...

CD22, also known as BL-CAM, is a type I transmembrane glycoprotein composed of two polypeptide chains, CD22alpha and CD22beta, with molecular weights of 130 and 140 kDa, respectively. These chains are produced by alternative splicing of the CD22 gene. CD22 is prominently expressed on mature B cells and B cell lymphomas, including hairy cell leukemia, diffuse large B-cell lymphoma, and nodular lymphocyte predominance Hodgkin's lymphoma, but is negative in classical Hodgkin's lymphoma. The extracellular portion of CD22 contains seven Ig-like domains that preferentially bind alpha2,6-linked sialic acid moieties found on epithelial, endothelial, B, and T cells. This binding can be masked by cis interactions with sialic acids on the same cell surface. CD22 expression is limited to late stages of B-cell differentiation, making it useful for phenotyping mature leukemias. Intracellularly, CD22 features six tyrosine residues within immunotyrosine-based inhibitory motifs (ITIM) and activation-like motifs. These residues are phosphorylated upon B-cell receptor engagement, allowing CD22 to regulate B-cell receptor signaling. CD22 participates in positive regulation through interactions with Src family tyrosine kinases and acts as an inhibitory receptor by recruiting cytoplasmic phosphatases via SH2 domains, which block signal transduction through dephosphorylation of signaling molecules. CD22's role in both positive and negative regulation of B-cell signaling, along with its specific expression pattern, makes it a valuable marker for antibody customers interested in B-cell-related research and diagnostics.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD22
Recombinant Monoclonal
1.34 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P20273
CD22
Recombinant fusion protein containing a sequence corresponding to amino acids 20-687 of human CD22 (NP_001762.2).
100 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
3F2M10
Unconjugated
CD22
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14
Rabbit
Protein A
RUO
933
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.