missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphA6 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149131
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
EPHA6 is a member of the ephrin receptor subfamily of protein-tyrosine kinases which have been implicated in axon guidance, neuron-target interactions, regional compartmentalization, and synaptic functions in nervous systems. EPHA6 is highly expressed in the brain and testis. Reduction in EPHA6 has been detected in Hypospadias, a common defect affecting the growth and closure of the external genitalia. Genetic inhibition of EPHA6 in mice produces behavioral deficits specifically in tests of learning and memory. Mice deficient in EPHA6 show reduced memory of the consequences of the training context.
Specifications
| EphA6 | |
| Polyclonal | |
| Unconjugated | |
| Epha6 | |
| DKFZp434C1418; EHK2; EHK-2; EK12; EPA6; EPH homology kinase 2; Eph receptor A6; EPHA6; EPH-like kinase 12; ephrin receptor EphA6; ephrin type-A receptor 6; FLJ35246; HEK12; m-ehk2; PRO57066; tyrosine-protein kinase receptor EHK-2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 13840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q62413 | |
| Epha6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA6, Ser28-Gln546. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction