Learn More
CD22 Mouse, anti-Human, PE, Clone: HIB22, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60083PE
This item is not returnable.
View return policy
Description
- The HIB22 antibody reacts with CD22 (Siglec-2), a type I transmembrane glycoprotein expressed in the cytoplasm of pro-B and pre-B cells and on the surface of mature and activated B cells and B cell lymphomas, but not on plasma cells
- CD22 functions as an inhibitory receptor for B cell receptor signaling and plays roles in modulating the activation threshold of B cells and controlling CD40 signaling
- CD22 comprises two isoforms generated by alternative splicing: CD22α (∼130 kDa) and CD22β (∼140 kDa)
- Monomeric CD22β is the most abundant isoform but an αβ heterodimer is also expressed
- CD22 is a member of the sialoadhesion subgroup within the immunoglobulin superfamily, with an N-terminal V-type Ig domain followed by six C2-type Ig domains
- Binding of the HIB22 antibody reportedly requires the presence of the two N-terminal-most domains
- Both the cytoplasmic and surface-bound forms of CD22 are recognized by the HIB22 antibody and its binding does not inhibit CD22-mediated cyto-adhesion
Specifications
| CD22 | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Chimpanzee, Human | |
| B Cells |
| Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| HIB22 | |
| Mouse | |
| Primary | |
| IgG1 κ |