missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen H053T02Y01-A
This item is not returnable.
View return policy
Description
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow Cytometry
Specifications
| CD22 | |
| Monoclonal | |
| 0.2 μg/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P20273 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| eBio4KB128 (4KB128) | |
| NovaFluor Yellow 570 | |
| CD22 | |
| A530093D23, B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, B-lymphocyte cell adhesion molecule (BL-CAM), Cd22, CD22 antigen, CD22 molecule, FLJ22814, Lectin 2, Leu-14, Lyb8, Lyb-8, MGC130020, sialic acid binding Ig-like lectin 2, sialic acid-binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2 (Siglec-2), Siglec2, Siglec-2, T-cell surface antigen Leu-14 | |
| Affinity chromatography | |
| RUO | |
| 933 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction