missing translation for 'onlineSavingsMsg'
Learn More

CD217 (IL-17Ra) Monoclonal Antibody (424LTS), Functional Grade, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-7917-050

 View more versions of this product

Product Code. 17171552

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The 424LTS monoclonal antibody reacts with and blocks IL-17A binding to human interleukin 17 receptor A (IL-17RA, IL-17R). IL-17RA is a type I membrane protein of approximately 96 kDa (866 amino acids) and is involved in inflammatory responses to IL-17A, IL-17F and IL-17AF heterodimer. It forms heterodimeric complexes with several other members of its receptor family, including IL-17RC for IL-17A and IL-17F signaling and IL-17RC for IL-17E (IL-25) signaling. IL-17RA is ubiquitously expressed, but in leukocytes its expression is elevated on granulocytes and monocytes. Applications Reported: This 424LTS antibody has been reported for use in blocking IL-17 receptor A ligand binding. Applications Tested: The ND50 of 424LTS as measured by the inhibition of IL-6 induction in NHDF cells is 1-5 μg/mL in the presence of 3 ng/mL of recombinant Human IL-17A. Neutralization dose will vary depending on assay method, cytokine concentration, and cell type. This antibody should be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

CD217 (IL-17Ra) Monoclonal antibody specifically detects CD217 (IL-17Ra) in Human samples. It is validated for Functional Assay, Neutralization
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD217 (IL-17Ra)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Q96F46
Mouse
50 mg
Primary
Human
Antibody
IgG1 κ
Functional Assay, Neutralization
424LTS
Functional Grade
Il17ra
AW538159; CANDF5; CD127; CD217; CDw217; hIL-17R; IL-17 receptor A; Il17r; Il17ra; IL-17RA; interleukin 17 receptor A; interleukin-17 receptor A; MGC10262; VDw217
Affinity chromatography
RUO
23765
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.