Learn More
CD210a Rat anti-Human, Brilliant Violet 421, Clone: 3F9, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 742942
Description
The 3F9 monoclonal antibody specifically binds to CD210a, which is also known as, Interleukin-10 Receptor subunit alpha (IL-10R subunit alpha/IL-10R /IL10RA), or Interleukin-10 receptor subunit 1 (IL-10R1). CD210a is a 90-110 kDa type I transmembrane glycoprotein that belongs to the type II cytokine receptor family. CD210a combines with IL-10 Receptor subunit beta (IL-10R /IL10RB/CDw210b) to form the IL-10 Receptor complex ( ) that can bind IL-10 and transduce signals intracellularly through the JAK/STAT pathway. IL-10 can suppress antigen presentation and the expression of proinflammatory type-1 immune responses while promoting type-2 immune responses. CD210a is expressed on T cells, B cells, NK cells, monocyte, macrophages and dendritic cells. Clone 3F9 is specific for human CD210a and its binding to the receptor can be blocked by recombinant human IL-10 (rhIL-10) protein.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| CD210a | |
| 3F9 | |
| Brilliant Violet 421 | |
| CD210, CDW210A, IL10RA, IL-10RA, IL-10R-A, IL10R, HIL-10R, IL-10R1, I10R1 | |
| Human IL-10R alpha Recombinant Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.