Learn More
CD210 (IL-10R) Monoclonal Antibody (1B1.3a), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-2101-38
Description
Description: The 1B1.3A monoclonal antibody reacts with mouse CD210, which is also known as the IL-10 receptor. CD210 is a member of the interferon receptor-like family that is expressed on a variety of hematopoietic cells, including thymocytes, T cells, B cells, NK cells, monocytes, and macrophages. Binding of IL-10 to its receptor initiates JAK/STAT signaling that leads to B cell proliferation and inhibition of macrophage activation. The 1B1.3A antibody has been reported to have neutralizing activity. Applications Reported: The 1B1.3A antibody has been reported for use in functional assays. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD210 (IL-10R) Monoclonal antibody specifically detects CD210 (IL-10R) in Mouse samples. It is validated for Functional Assay
Specifications
| CD210 (IL-10R) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q61727 | |
| Rat | |
| 5 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Functional Assay | |
| 1B1.3a | |
| Functional Grade | |
| Il10ra | |
| AW553859; CD210; CD210a; CDw210; CDW210A; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; HIL-10R; I10R; I10R1; il-10 receptor; il-10 receptor antagonist; IL-10 receptor subunit alpha; IL10R; IL-10R alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10R1; IL-10R2; IL10RA; IL-10RA; interleukin 10 receptor subunit alpha; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha; mIL-10R | |
| Affinity chromatography | |
| RUO | |
| 16154 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.