Learn More
CD21 Mouse anti-Human,Baboon,Cynomolgus,Pig,Rhesus Ma, BB700, Clone: B-ly4, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566569
 View more versions of this product
Product Code. 15815159
  • ONLINE EXCLUSIVE PRICE
    £370.00
    £354.98 / Each
    Save £15.02
    4% Off
Description
CD21 Monoclonal antibody specifically detects CD21 in Human,Baboon,Cynomolgus,Pig,Rhesus Ma samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD21
Monoclonal
Brilliant Blue 700 (BB700)
100 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Flow Cytometry
B-ly4
Mouse
RUO
Human, Baboon, Cynomolgus, Pig, Rhesus Macaque
Purified