missing translation for 'onlineSavingsMsg'
Learn More

CD21/CD35 Antibody (7E9), Novus Biologicals™

Product Code. 18377521 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377521 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377521 Supplier Novus Biologicals Supplier No. NBP311263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD21/CD35 Monoclonal specifically detects CD21/CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD21/CD35
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 7000000000
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias C3b/C4b receptor, C3-binding protein, C3BR, C3DR, C4BR, CD35, CD35 antigen, complement component (3b/4b) receptor 1 (Knops blood group), complement component (3d/Epstein Barr virus) receptor 2, complement component 3d receptor 2, complement receptor 1, complement receptor type 1, complement receptor type 2, CR, CVID7, EBV receptor, KN, Knops blood group antigen, SLEB9
Host Species Rat
Immunogen Purified mouse CR1 from serum
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.