missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (CR2/1952), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60733AF700
This item is not returnable.
View return policy
Description
CD21 Monoclonal specifically detects CD21 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD21 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CR2 | |
| Recombinant fragment (around aa 44-196) of human CR2 (CD21) protein (exact sequence is proprietary) (Uniprot: P20023) | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology | |
| Recognizes a protein of 140kDa, which is identified as the complement receptor 2 (CR2)/ CD21. This protein is expressed strongly on mature B cells, follicular dendritic cells and weakly on immature thymocytes and T lymphocytes. In B-cell ontogeny, CD21 appears after the pre-B-stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD21 expression is also gradually lost after stimulation of B cells in vitro. CD21 functions as receptor for C3d, C3dg and iC3b Complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CR2/1952 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction