missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD209b (SIGN-R1) Armenian Hamster anti-Mouse, Clone: eBio22D1 (22D1), eBioscience™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen 14-2093-81
This item is not returnable.
View return policy
Description
The eBio22D1 monoclonal antibody reacts with mouse SIGNR1 (CD209b). SIGNR1 is a type II transmembrane C-type lectin that was identified in a search for mouse homologues of human DC-SIGN. It is expressed at high levels in splenic marginal zone macrophages and lymph node medullary macrophages, where it functions to uptake dextran polysaccharides, including the capsular polysaccharide of Streptococcus pneumoniae. It has also been demonstrated that SIGNR1 physically associates with TLR4/MD2, and it has been suggested that this association plays a role in recognition of LPS. Furthermore, recently it has been shown that SIGNR1 deficient mice have a defect in catabolism of the complement component C3, and that SIGNR1 binds directly to the complement C1 subcomponent, C1q to assemble a non-conventional C3 convertase. The eBio22D1 monoclonal antibody does not cross-react with the closely related SIGNR1, SIGNR2, SIGNR3 or SIGNR4.Specifications
CD209b (SIGN-R1) | |
Monoclonal | |
0.5mg/mL | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
Armenian Hamster | |
RUO | |
Mouse | |
Purified |
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation | |
eBio22D1 (22D1) | |
Unconjugated | |
C-type lectin, SIGNR1 | |
50 μg | |
Primary | |
Store at 2-8°C. | |
IgG |
For Research Use Only.