missing translation for 'onlineSavingsMsg'
Learn More

CD209a Monoclonal Antibody (MMD3), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-2094-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15550207

  • £407.00 / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This MMD3 monoclonal antibody recognizes mouse CD209a, which is also known as DC-SIGN. CD209a is a type II transmembrane C-type lectin expressed on a subset of dendritic cells, including some CD4+, CD8- and plasmacytoid pre-dendritic cells. Studies indicate that CD209a expression can vary according to the activation state of the host. Moreover, CD209a is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. CD209a is involved in mediating the innate immune response by binding microbial carbohydrates. Cross-blocking studies suggest that MMD3 recognizes a different a different epitope from LWC06 and 5H10. Applications Reported: This MMD3 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, microscopy, immunohistochemical staining, and immunocytochemistry. Applications Tested: This MMD3 antibody has been tested by immunohistochemistry on frozen 5-week-old mouse thymus and can be used at less than or equal to 20 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants.
Specifications

Specifications

CD209a
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q91ZX1
Cd209a
Affinity chromatography
RUO
170786
4° C, store in dark, DO NOT FREEZE!
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen)
MMD3
Alexa Fluor 488
Cd209a
CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5
Mouse
100 μg
Primary
Mouse
Antibody
IgG2c κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.