missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD208 (DC-LAMP) Monoclonal Antibody (31B), APC, eBioscience™
GREENER_CHOICE

Product Code. 15558056
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15558056 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558056 Supplier Invitrogen™ Supplier No. 17208941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: This 31B monoclonal antibody reacts with human CD208, which is also known as dendritic cell lysosome-associated membrane glycoprotein (DC-LAMP). CD208 is expressed almost exclusively by mature dendritic cells. Expression of this molecule can be induced in monocytes by stimulation with GM-CSF and IL-4. Moreover, CD208 can be upregulated in CD34+-derived dendritic cells by GM-CSF, SCF, and TNFa. CD208 is also expressed in dendritic cells of non-lymphoid tissues such as skin. While CD208 can be detected on the cell surface, this protein is mainly localized within lysosomes. CD208 plays a key role in the processing and presentation of antigen during the immune response. The 31B antibody only recognizes intracellular CD208, not the cell surface-expressed protein. Applications Reported: This 31B antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 31B antibody has been pre-titrated and tested by flow cytometric analysis of in vitro differentiated mature dendritic cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Dendritic cells (DCs) are the most potent antigen-presenting cells. Immature DCs efficiently capture antigens and differentiate into interdigitating dendritic cells (IDCs) in lymphoid tissues that induce primary T-cell responses (summary by de Saint-Vis et al., 1998 [PubMed 9768752]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD208 (DC-LAMP)
Applications Flow Cytometry
Classification Monoclonal
Clone 31B
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene LAMP3
Gene Accession No. Q9UQV4
Gene Alias 1200002D17Rik; CD208; DC LAMP; DCLAMP; DC-LAMP; DC-lysosome-associated membrane glycoprotein; dendritic cells-lysosome associated membrane glycoprotein; LAMP; Lamp3; LAMP-3; lysosomal associated membrane protein 3; lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; Protein TSC403; TSC403
Gene Symbols LAMP3
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27074
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.