missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18704-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes the intracellular carboxy-terminal part of CD206 and MRC1L1.
CD206 (macrophage mannose receptor, MMR), also known as mannose receptor C1 (MRC1), is a type I transmembrane glycoprotein serving as pattern recognition receptor for carbohydrate groups on the surface of bacteria, fungi and other pathogens. Expressed mainly on tissue macrophages and dendritic cells, CD206 mediates endocytosis of these pathogens and presentation of their antigens to the adaptive immune system. CD206 can also be detected in a soluble form in human plasma and is elevated in patients with acute sepsis.Specifications
| CD206 | |
| Polyclonal | |
| Unconjugated | |
| MRC1 | |
| CD206, CLEC13D, Macrophage mannose receptor 1, Mannose receptor, mannose receptor, C type 1, MMR, MRC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17533, 291327, 4360 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22897, Q61830 | |
| Mrc1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction