missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 (MMR) Monoclonal Antibody (MR6F3), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen 56-2061-80
This item is not returnable.
View return policy
Description
Rat Monoclonal Antibody
CD206 (MMR) Monoclonal antibody specifically detects CD206 (MMR) in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD206 (MMR) | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| Q61830 | |
| Mrc1 | |
| Affinity chromatography | |
| RUO | |
| 17533 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| MR6F3 | |
| Alexa Fluor 700 | |
| Mrc1 | |
| AW259686; bA541I19.1; CD206; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; hMR; Human mannose receptor; Macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; MMR; MR; Mrc1; MRC1L1 | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction