missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD205 Monoclonal Antibody (205yekta), NovaFluor™ Red 700, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

£160.00 - £300.00

Specifications

Antigen CD205
Clone 205yekta
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055468
View Documents
Invitrogen™
M051T03R03-A
100 μg
£300.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30054925
View Documents
Invitrogen™
M051T02R03-A
25 μg
£160.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD205 Monoclonal antibody specifically detects CD205 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD205
0.1 mg/mL
Monoclonal
Rat
Mouse
Q60767
17076
Primary
4°C, store in dark, DO NOT FREEZE!
205yekta
Flow Cytometry
NovaFluor Red 700
RUO
PBS with 0.09% sodium azide; pH 7.2
CA-IX; CD205; CLEC13B; C-type lectin domain family 13 member B; DCL-1 fusion; DEC205; DEC-205; DEC-205/CD205 protein; gp200; gp200-MR6; group VI C-type animal lectin; LY75; ly-75; Lymphocyte antigen 75; membrane antigen MN; P54/58N; pMW1; RGD1565962; RP23-10L17.4
IgG2a κ
Affinity chromatography
LY75
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.