missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD204 Monoclonal Antibody (M204PA), PE-Cyanine7, eBioscience™, Invitrogen™

Product Code. 16380764
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16380764 25 μg 25µg
16390764 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16380764 Supplier Invitrogen™ Supplier No. 25204680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

This M204PA antibody recognizes mouse CD204 also known as Macrophage Scavenger Receptor 1 (MSR 1), Scavenger Receptor A (SR-A) or SCARA1. CD204 belongs to the class A family of scavenger receptors that are primarily responsible for binding a wide variety of macromolecules and are involved in many aspects of macrophage activity. Other members of this family include: MARCO (SCARA2), CSR (SCARA3), SRCL (SCARA4) and SCARA5. All class A scavenger receptors are trans-membrane proteins with extracellular C-termini and homotrimeric structures. CD204 exists as 2 splice variants: the more ubiquitous full length isoform (type I) and a shorter isoform (type II) that lacks the C-terminal cysteine-rich domain. Both types are functional and bind modified LDL with similar affinity. In addition to lipid metabolism, CD204 plays an important role suppressing inflammation and macrophage activation. CD204 is considered one of the markers of M2 and Tumor-Associated Macrophages. Interestingly, this molecule is responsible for adherence of RAW264.7 cells to tissue culture-treated plastic in the absence of Ca and Mg cations. This M204PA antibody binds to CD204 of multiple mouse strains, including C57Bl/6, Balb/c and Swiss-Webster (outbred). This antibody works on fixed cells and can be used to stain CD204 intracellularly. The M204PA antibody does not cross-react with human or rat. This antibody is not suitable for IHC staining.

CD204 gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are macrophage-specific trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes including atherosclerosis, Alzheimer's disease, and host defense. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. It has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a dominant negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. Diseases associated with MSR1 include Barrett Esophagus and Prostate Cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD204
Applications Flow Cytometry
Classification Monoclonal
Clone M204PA
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide
Gene MSR1
Gene Accession No. P30204
Gene Alias bovine scavenger receptor type II; CD204; Macrophage acetylated LDL receptor I and II; macrophage scavenger receptor 1; macrophage scavenger receptor type III; macrophage scavenger receptor types I and II; MRS-A; MSR; Msr1; MSR-A; phSR1; phSR2; RGD1564316; SCARA1; Scavenger receptor class A member 1; scavenger receptor class A, member 1; scavenger receptor type A; scavenger receptor type I; Scvr; SRA; SR-A; SR-AI; SR-AI/II; SR-AII
Gene Symbols MSR1
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20288
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.