missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD202b (TIE2) Polyclonal Antibody

CD202b (TIE2) Polyclonal Antibody
Brand: Thermo Scientific PA5116042
This item is not returnable.
View return policy
Description
CD202b (TIE2) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| CD202b (TIE2) | |
| Polyclonal | |
| Unconjugated | |
| TEK | |
| AA517024; Angiopoietin-1 receptor; CD202B; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; Tie2; tie-2; tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21687, 7010 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q02763, Q02858 | |
| TEK | |
| A synthesized peptide derived from human TEK(Accession Q02763), corresponding to amino acid residues E872-T922. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction