Learn More
Invitrogen™ CD201 (EPCR) Monoclonal Antibody (RCR-227), APC, eBioscience™
Rat Monoclonal Antibody
£244.00
Specifications
| Antigen | CD201 (EPCR) |
|---|---|
| Clone | RCR-227 |
| Concentration | 5 μL/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: The monoclonal antibody RCR-227 reacts with human CD201, which is also known as the endothelial protein C receptor (EPCR), a 46-kDa transmembrane protein related to class I MHC. CD201 was originally identified on vascular endothelium cells, predominantly larger blood vessels. It is also expressed on monocytes, lymphocytes, hematopoietic stem cells, neutrophils, and eosinophils. CD201 serves as a binding site for protein C, activated protein C (APC), and factor VII/VIIa. A soluble form has also been identified in the plasma. CD201 plays a role in controlling coagulation as disruption of its gene in mice caused placental thrombosis and early embryonic death. This cell surface molecule is also involved in the anti-inflammatory response. APC binding to CD201 enables PAR-1 activation that confers cytoprotective effects. In monocytes, APC binding decreases secretion of proinflammatory cytokines, chemokines, and growth factors. Finally, CD201 has been shown to inhibit lymphocyte migration. Applications Reported: This RCR-227 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RCR-227 antibody has been pre-titrated and tested by flow cytometric analysis of HUVECs. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-...
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.Specifications
| CD201 (EPCR) | |
| 5 μL/Test | |
| Flow Cytometry | |
| APC | |
| Rat | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q9UNN8 | |
| 10544 | |
| Primary | |
| Affinity chromatography |
| RCR-227 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| Procr | |
| Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
| Procr | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.