missing translation for 'onlineSavingsMsg'
Learn More

CD200R3 Antibody (Ba91), Novus Biologicals™

Product Code. 18359091 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359091 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359091 Supplier Novus Biologicals Supplier No. NBP311253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD200R3 Monoclonal specifically detects CD200R3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R3
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone Ba91
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunoprecipitation, Functional
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias CD200 cell surface glycoprotein receptor-like b, CD200 receptor-like 3, CD200RLb, Cell surface glycoprotein CD200 receptor 3, Cell surface glycoprotein OX2 receptor 3
Host Species Rat
Immunogen Mouse primary basophils; cell-enriched bone marrow cells
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74603
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.