missing translation for 'onlineSavingsMsg'
Learn More

CD200R1L Antibody, DyLight 594, Novus Biologicals™

Product Code. 18393573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393573 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393573 Supplier Novus Biologicals Supplier No. NBP306395DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD200R1L Polyclonal antibody specifically detects CD200R1L in Cynomolgus Monkey samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R1L
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cynomolgus CD200R1L (Met 1-Leu 240).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344807
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.