missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R1 Antibody (380525), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3414MFV450
This item is not returnable.
View return policy
Description
CD200R1 Monoclonal antibody specifically detects CD200R1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CD200R1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 380525 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
| Mouse myeloma cell line NS0-derived recombinant human CD200 R1, Ala27-Leu266 (predicted), Accession # NP_620161 | |
| 0.1 mL | |
| Cancer, Neuronal Cell Markers, Neuroscience | |
| 131450 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction