missing translation for 'onlineSavingsMsg'
Learn More

CD200R1 Antibody (019), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18379000 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379000 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379000 Supplier Novus Biologicals Supplier No. NBP290481AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD200R1 Monoclonal specifically detects CD200R1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R1
Applications ELISA
Classification Monoclonal
Clone 19
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200R1 (NP_067300.1; Met 1-Pro 238).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 131450
Test Specificity No cross-reactivity in ELISA with: Human CD200R1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.