Learn More
Invitrogen™ CD200R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596991
Description
Immunogen sequence: SLYIELLPVP GAKKSAKLYI PYIILTIIIL TIVGFIWLLK VNGCRKYKLN KTESTPVVEE DEMQPYASYT EKNNPLYDTT NKVKASEALQ SEVDTDLHTL; Positive Samples: U-937, NCI-H460; Cellular Location: Cell membrane, Single-pass type I membrane protein, Single-pass type I membrane protein.
CD200R (OX2R) is a receptor for the OX-2 membrane glycoprotein, and both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. CD200R is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. In mouse, CD200R is expressed primarily by peripheral blood monocytes and neutophils but also by other leucocytes including T-lymphocytes and mast cells. Further mouse studies of a gene related to CD200R suggests that the receptor may control myeloid function in a tissue-specific manner. Alternative splicing of CD200R results in multiple transcript variants. Diseases associated with CD200R dysfunction include primary hypertrophic osteoarthropathy.
Specifications
| CD200R | |
| Polyclonal | |
| Unconjugated | |
| CD200R1 | |
| antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 131450 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TD46 | |
| CD200R1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 249-348 of human CD200R1 (NP_6201611). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.