missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200/OX2 Antibody (OX-104), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
£428.00
Specifications
| Antigen | CD200/OX2 |
|---|---|
| Clone | OX-104 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
Description
CD200/OX2 Monoclonal antibody specifically detects CD200/OX2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| CD200/OX2 | |
| Flow Cytometry | |
| FITC | |
| Mouse | |
| Immunology | |
| PBS | |
| 4345 | |
| IgG1 κ | |
| Protein A purified |
| OX-104 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
| Made to Human CD200 | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title