missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200/OX2 Antibody (OX-102), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65603APC
This item is not returnable.
View return policy
Description
CD200/OX2 Monoclonal specifically detects CD200/OX2 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD200/OX2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CD200 | |
| Membrane fraction of thioglycollate-elicited rat peripheral cells. | |
| 0.125 mL | |
| Immunology | |
| 4345 | |
| Rat | |
| Purified |
| Flow Cytometry | |
| OX-102 | |
| Flow Cytometry | |
| antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65603 recognizes the rat OX2 (CD200) receptor 1. This antigen is a heavily glycosylated 60-100kD cell surface molecule expressed by cells of the myeloid lineage but not by T or B lymphocytes. This antibody has been shown to block the interaction of OX2 receptor 1 with CD200. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction