missing translation for 'onlineSavingsMsg'
Learn More

CD200/OX2 Antibody (333), FITC, Novus Biologicals™

Product Code. 18360701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360701 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360701 Supplier Novus Biologicals Supplier No. NBP289696F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200/OX2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 333
Conjugate FITC
Formulation PBS
Gene Alias antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD200/OX2 (NP_005935.4; Met1-Gly232).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity Has cross-reactivity in ELISA with: Mouse CD200
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.