missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD200/OX2 Monoclonal specifically detects CD200/OX2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | CD200/OX2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2 |
| Conjugate | Biotin |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200/OX2 (NP_001076017.1; Met 1-Gly 232). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?