missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200 Mouse anti-Human, Clone: 5F3D6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66282-1-IG-150UL
This item is not returnable.
View return policy
Description
CD200 is a type-1 membrane glycoprotein that contains two immunoglobulin domains, and belongs to the immunoglobulin superfamily. CD200 is a broadly distributed cell surface glycoprotein that interacts with a receptor on myeloid cells (CD200R) and is implicated in locally preventing macrophage activation. Distant, but recognizable, homologues of CD200 have been identified in many herpesviruses and poxviruses. Studies of the related genes in mouse and rat suggest that CD200 may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms of CD200 have been found. Diseases associated with CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy.Specifications
| CD200 | |
| Monoclonal | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P41217 | |
| CD200 | |
| CD200 Fusion Protein Ag5045 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 5F3D6 | |
| Unconjugated | |
| CD200 | |
| CD200, CD200 molecule, MOX1, MOX2, MRC, OX 2, OX 2 membrane glycoprotein | |
| Mouse | |
| Protein G | |
| RUO | |
| 4345 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction